MABN779
Anti-Amyloid Beta 3NTyr10 Antibody, clone 4A5E8
clone 4A5E8, from mouse
Synonym(s):
Ab42, Amyloid beta 1-42, Abeta42, Abeta42(3NTyr10), Abeta42 (3NY10)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
4A5E8, monoclonal
species reactivity
human
technique(s)
ELISA: suitable
immunohistochemistry: suitable
isotype
IgG2bκ
NCBI accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... APP(351)
General description
Specificity
Immunogen
Application
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Amyloid Beta 3NTyr10 in unfixed cortical brain sections from a control verses clinically confirmed AD (Braak stage V) cases. (Courtesy of Dr. Markus Kummer and Prof. Dr. Michael Heneka, MD, University of Bonn, Department of Neurology, Clinical Neurosciences, Germany).
Neuroscience
Quality
Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG2b .
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service