Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
41106514
NACRES:
NA.76
eCl@ss:
32011203
Product HTS045RTA is not currently sold in your country. Contact Technical Service
Quality Segment
description
Cell line type: GPCR Cell Lines, GPCR Class A, protein target OX2
technique(s)
calcium flux assay: suitable
detection method
fluorometric
General description
Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.
The orexin/hypocretin peptides orexin A and orexin B (also known as hcrt-1 and hcrt-2) are 33- and 28-amino acid peptides, respectively, that are preferentially expressed in hypothalamus. The orexins bind to two GPCRs, OX1 and OX2, to increase intracellular calcium. Orexins function to increase wakefulness, primarily through OX2, which has been shown to harbor the causative mutation in narcoleptic dogs (Lin et al., 1999). In addition, the orexinergic system plays a role in feeding and stress responses (Kukkonen et al., 2002). Millipore′s cloned human OX2-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant OX2 expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at OX2 Receptor.
The orexin/hypocretin peptides orexin A and orexin B (also known as hcrt-1 and hcrt-2) are 33- and 28-amino acid peptides, respectively, that are preferentially expressed in hypothalamus. The orexins bind to two GPCRs, OX1 and OX2, to increase intracellular calcium. Orexins function to increase wakefulness, primarily through OX2, which has been shown to harbor the causative mutation in narcoleptic dogs (Lin et al., 1999). In addition, the orexinergic system plays a role in feeding and stress responses (Kukkonen et al., 2002). Millipore′s cloned human OX2-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant OX2 expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at OX2 Receptor.
Other Notes
Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.
Disclaimer
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.