Skip to Content
Merck
All Photos(1)

Key Documents

EZCURVSCR

Millipore

Membrane exposure adjustment screwdriver

General microbiological analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

Quality Level

manufacturer/tradename

EZ-Pak®

input

sample type bottled water
sample type cosmetics
sample type beer
sample type pharmaceutical(s)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

General description

EZ-Pak Spare Part

Application

General microbiological analysis

Legal Information

EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service