Skip to Content
Merck
All Photos(1)

Key Documents

ABE473

Sigma-Aldrich

Anti-hRNF20 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

E3 ubiquitin-protein ligase BRE1A, BRE1-A, hBRE1, RING finger protein 20, hRNF20

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
3 050,00 kr

3 050,00 kr


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
3 050,00 kr

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

3 050,00 kr


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RNF20(56254)

General description

BRE1-A (E3 ubiquitin-protein ligase BRE1A) also known by it′s gene name hRNF20, plays a role in histone code and gene regulation as a component of the RNF20/40 E3 ubiquitin-protein ligase complex. That complex mediates monoubiquitination of ′Lys-120′ of histone H2B,which is a specific tag for epigenetic transcriptional activation as well as a prerequisite for histone H3 ′Lys-4′ (H3K4me) and ′Lys-79′ (H3K79me) methylation.

Immunogen

GST-tagged recombinant protein corresponding to human hRNF20.

Application

This hRNF20 antibody is validated for use in WB for the detection of the hRNF20 protein.

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected hRNF20 in 10 µg of A431 cell lysate.

Target description

~113 kDa observed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service