Skip to Content
Merck
All Photos(1)

Key Documents

4.90288

Sigma-Aldrich

1-Butyl-2,3-dimethylimidazolium bis(trifluoromethylsulfonyl)imide

for synthesis

Synonym(s):

1-Butyl-2,3-dimethylimidazolium bis(trifluoromethylsulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17F6N3O4S2
CAS Number:
Molecular Weight:
433.39
MDL number:
UNSPSC Code:
12352107
form:
liquid
Assay:
≥98.0% (HPLC)
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

form

liquid

potency

50-300 mg/kg LD50, oral (Rat)

density

1.437 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H17N2.C2F6NO4S2/c1-4-5-6-11-8-7-10(3)9(11)2;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h7-8H,4-6H2,1-3H3;/q+1;-1

InChI key

UCCKRVYTJPMHRO-UHFFFAOYSA-N

Analysis Note

Purity (HPLC, area%): ≥ 98.0 %
Identity (NMR): passes test
Halides (IC): ≤ 0.1 %
Water (K. F.): ≤ 1.0 %

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service