Mer RTK Inhibitor, UNC569, 1350547-65-7 is a potent, reversible and ATP-competitive inhibitor of Mer receptor tyrosine kinase (IC50 = 2.9 nM; Ki = 4.3 nM).
Synonym(s):
Mer RTK Inhibitor, UNC569, MERTK Inhibitor I, RP38 Inhibitor I, TAM Family RTK Inhibitor I, UNC-569, 1-(( trans-4-Aminocyclohexyl)methyl)-N-butyl-3-(4-fluorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-6-amine, MERTK Inhibitor I, RP38 Inhibitor I, TAM Family RTK Inhibitor I, UNC-569, 1-((trans-4-Aminocyclohexyl)methyl)-N-butyl-3-(4-fluorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-6-amine
An orally bioavailable pyrazolopyrimidine compound that acts as a potent, reversible and ATP-competitive inhibitor of Mer receptor tyrosine kinase (IC50 = 2.9 nM; Ki = 4.3 nM) with ~10-fold greater selectivity over other TAM family kinases (IC50 = 37 and 48 nM for Axl and Tyro3, respectively). Also shown to affect the activities of Flt3, MAPKAPK2, RET and Ret-Y791F at 30 nM by 82%, 92%, 59% and 56% in a 72 kinase panel, and block Mer auto-phosphorylation in human Pre-B leukemia 697 cells with an IC50 of 141 nM. Exhibits desirable pharmacokinetic profile and efficacy in mice.
Warning
Toxicity: Standard Handling (A)
Physical form
Supplied as a trifluoroacetate salt.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable fo rup to 6 months at -20°C.
Other Notes
Liu, J., et al. 2012. ACS Med. Chem. Lett.3, 129.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.