Skip to Content
Merck
All Photos(1)

Key Documents

119107

Sigma-Aldrich

ATP Assay Kit

Synonym(s):

Luciferin-Related Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133

usage

sufficient for 200 tests

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

input

sample type soil
sample type blood
sample type urine
sample type milk
sample type sludge

detection method

chemiluminescent

storage temp.

−70°C

General description

Cell death (especially apoptosis) is an energy-dependent process that requires ATP. As ATP levels fall to a point where the cell can no longer perform basic metabolic functions, the cell will die. A typical apoptotic cell exhibits a significant decrease in ATP level. Therefore, loss of ATP level in the cell has been used as an indicator of cell death. In contrast, increased levels of ATP has been an indicator of cell proliferation. Calbiochem® brand ATP Assay Kit utilizes bioluminescent detection of the ATP levels for a rapid screening of apoptosis and cell proliferation simultaneously in mammalian cells. The assay utilizes luciferase to catalyze the formation of light from ATP and luciferin, and the light can be measured using a luminometer or Beta Counter.
The assay is based on the firefly luciferase-catalyzed oxidation of D-luciferin in the presence of ATP and oxygen, whereby the amount of ATP is quantified by the amount of light (hv) produced.

Components

One vial of Luciferase ATP Monitoring Enzyme, Enzyme Reconstitution Buffer, 1 bottle of Nucleotide Releasing Reagent, one vial of ATP Standard, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Specifications

Assay Time: 1 h

Principle

The assay can be fully automatic for high throughput (10 s/sample) and is extremely sensitive (detects 10-100 mammalian cells/well). The high sensitivity of this assay has led to many other applications for detecting ATP production in various enzymatic reactions, as well as for detecting low level bacterial contamination in samples such as blood, milk, urine, soil, and sludge.

Preparation Note

Reagent Reconstitution1. Reconstitute ATP Monitoring Enzyme with 2ml of the Enzyme Reconstitution Buffer. Mix well by gentle pipetting. The reconstituted enzyme is stable for up to 2 months at 4°C.2. Prepare an ATP standard solution by dissolving the 1 mg ATP into 1 ml of H2O. The solution is stable for several weeks at -20°C.

Storage and Stability

Upon arrival store the entire contents of the kit at -70°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Harshita Negi et al.
The Journal of general and applied microbiology, 59(1), 83-88 (2013-03-23)
In view of antibiotic resistance among pathogens, the present study is to address the toxicity of Ag2O nanoparticles against the Gram-positive and Gram-negative bacteria through in vitro assays. The preliminary screening by agar diffusion assay confirms the antibacterial activity of
Mariya A Borisova et al.
Scientific reports, 10(1), 21135-21135 (2020-12-05)
The disruption of the protective intestinal barrier-the 'leaky gut'-is a common complication of the inflammatory bowel disease. There is limited data on the mechanisms of the intestinal barrier disruption upon low-grade inflammation characteristic of patients with inflammatory bowel disease in
Yu-Chien Hung et al.
Cell reports, 36(12), 109729-109729 (2021-09-23)
Human ubiquinol-cytochrome c reductase core protein 1 (UQCRC1) is an evolutionarily conserved core subunit of mitochondrial respiratory chain complex III. We recently identified the disease-associated variants of UQCRC1 from patients with familial parkinsonism, but its function remains unclear. Here we
Bin Li et al.
Infection and drug resistance, 15, 961-973 (2022-03-19)
Hepatitis B virus (HBV) infection is one of the global public problems. Among the known infection cases, HBV X protein (HBx) is one of the key inducements of viral replication and host infection. This study was aimed to uncover the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service