Skip to Content
Merck
All Photos(1)

Key Documents

1.09727

Supelco

Polyethylene glycol

4000, for gas chromatography

Synonym(s):

Poly(ethylene glycol), Polyethylene glycol 4000, Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 4000, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

>320 °C (DIN 51794)

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

5-7 (20 °C, 100 g/L in H2O)

mp

53-58 °C

transition temp

flash point 270 °C

solubility

500 g/L

density

1.2 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

storage temp.

no temp limit

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: 9727-2; 9727

Analysis Note

Thermogravimetry (thermogravimetric): 2 % lost of weight between 320-390°C
McReynolds constant (Benzene): Mc-Renolds-Konstante: ca. 320
McReynolds constant (Butanol): Mc-Renolds-Konstante 1-Butanol: ca.550
McReynolds constant (Nitropropane): Mc-Renolds-Konstante: ca. 570

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service