Skip to Content
Merck
All Photos(3)

Documents

06-1091

Sigma-Aldrich

Anti-IL-13 receptor Antibody, alpha-2 (IL-13RA2)

from rabbit, purified by affinity chromatography

Synonym(s):

interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, interleukin 13 receptor alpha 2 chain, Interleukin-13-binding protein, IL-13R subunit alpha-2, cancer/testis antigen 19, IL-13 receptor subunit alpha-2, interleukin 13 binding pro

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL13RA2(3598)

General description

Interleukin-13 receptor subunit alpha-2 (IL-13RA2) is one receptor of Interleukin-13 which is a key moderator of asthma and allergic inflammation. IL-13 and its receptors have been shown to play a part in alcohol-mediated activation of pro-fibrotic pathways. IL-13 has a suppressive effect on macrophage activation but cannot activate T cells.

Specificity

This antibody recognizes Interleukin-13 receptor, alpha-2 at the cytoplasmic domain.

Immunogen

Epitope: Cytoplasmic Domain
KLH-conjugated linear peptide corresponding to human Interleukin-13 receptor, alpha-2 at the cytoplasmic domain.

Application

Anti-IL-13 receptor Antibody, alpha-2 (IL-13RA2) is an antibody against IL-13 receptor for use in WB, IH(P).
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
Western Blot (SNAP i.d. Protein Detection System) Analysis: 5 µg/mL from a previous lot detected Interleukin-13 receptor, alpha-2 in 10 µg of HeLa cell lysate.

Immunohistochemistry (paraffin) Analysis: A 1:500 dilution from a previous lot was used to detect Interleukin-13 receptor, alpha-2 in colorectal carcinoma and normal human kidney/medulla tissue sections.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Interleukin-13 receptor, alpha-2 in 10 µg of HeLa cell lysate.

Target description

~44 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Zhou et al.
The Journal of clinical investigation, 125(8), 3178-3192 (2015-06-30)
Hermansky-Pudlak syndrome (HPS) comprises a group of inherited disorders caused by mutations that alter the function of lysosome-related organelles. Pulmonary fibrosis is the major cause of morbidity and mortality in patients with subtypes HPS-1 and HPS-4, which both result from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service