Skip to Content
Merck
All Photos(1)

Key Documents

KEY001150963

Key Organics / BIONET

2,2,2-trifluoro-1-(1-methyl-1H-indol-3-yl)ethan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
229.2
MDL number:

mp

72 - 73

Iupac Name

2,2,2-trifluoro-1-(1-methyl-1H-indol-3-yl)ethanol

Inchi Code

1S/C11H10F3NO/c1-15-6-8(10(16)11(12,13)14)7-4-2-3-5-9(7)15/h2-6,10,16H,1H3

InChI key

ARPIRHUPDYIDHC-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/7R-1061

Melting Point

72 - 73

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary statements

MSDS

https://www.keyorganics.net/sds/7R-1061


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library