Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249D771

J & W Pharmlab LLC

4-Acetyl-3,4-dihydro-2H-benzo[1,4]oxazine-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
202.21
MDL number:

Iupac Name

4-acetyl-3,4-dihydro-2H-benzo[b][1,4]oxazine-3-carbonitrile

Inchi Code

1S/C11H10N2O2/c1-8(14)13-9(6-12)7-15-11-5-3-2-4-10(11)13/h2-5,9H,7H2,1H3

InChI key

SXMSPPHEQTXVSO-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Yellow Solid

Product Link

https://www.jwpharmlab.com/home/product/53R0251

MSDS

https://www.jwpharmlab.com/home/product/53R0251


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library