Skip to Content
Merck
All Photos(1)

Key Documents

ENAH3050302D

Enamine

3-(2-ethoxyphenyl)prop-2-enal

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
176.22

Iupac Name

(2E)-3-(2-ethoxyphenyl)-2-propenal

Inchi Code

1S/C11H12O2/c1-2-13-11-8-4-3-6-10(11)7-5-9-12/h3-9H,2H2,1H3/b7-5+

InChI key

QSZJUEOFIBFGPM-FNORWQNLSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1851021

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library