Skip to Content
Merck
All Photos(1)

Key Documents

ENA458437344

Enamine

5-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
148.21
MDL number:

mp

102-103

Iupac Name

5-methyl-1,2,3,4-tetrahydro[1,8]naphthyridine

Inchi Code

1S/C9H12N2/c1-7-4-6-11-9-8(7)3-2-5-10-9/h4,6H,2-3,5H2,1H3,(H,10,11)

InChI key

BENBACKRAYKPOZ-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-239138

Melting Point

102-103

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-239138


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library