ENA341296031
Enamine
5-(4-nitrophenyl)-1,3-oxazole-4-carboxylic acid
About This Item
Iupac Name
5-(4-nitrophenyl)-1,3-oxazole-4-carboxylic acid
Inchi Code
1S/C10H6N2O5/c13-10(14)8-9(17-5-11-8)6-1-3-7(4-2-6)12(15)16/h1-5H,(H,13,14)
InChI key
PUWYNVGKYCYJJF-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-92732
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.