Skip to Content
Merck
All Photos(1)

Key Documents

CH6447916643

Chem Impex International, Inc.

Glycine 4-methoxy-b-naphthylamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
266.73
MDL number:

Iupac Name

2-amino-N-(4-methoxy-2-naphthyl)acetamide hydrochloride

Inchi Code

1S/C13H14N2O2.ClH/c1-17-12-7-10(15-13(16)8-14)6-9-4-2-3-5-11(9)12;/h2-7H,8,14H2,1H3,(H,15,16);1H

InChI key

OPTGKLWGIASWQF-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

99%

Country of Origin

US

Shipping Temperature

RT

Physical Form

White powder

Product Link

https://www.chemimpex.com/product/productinfo/0/3974

MSDS

https://www.chemimpex.com/product/productinfo/0/3974


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library