CH6371387271
Chem Impex International, Inc.
5-(2,4-Dichlorophenyl)isoxazole-3-carboxylic acid
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
mp
178-186C
Iupac Name
5-(2,4-dichlorophenyl)-3-isoxazolecarboxylic acid
Inchi Code
1S/C10H5Cl2NO3/c11-5-1-2-6(7(12)3-5)9-4-8(10(14)15)13-16-9/h1-4H,(H,14,15)
InChI key
JTPPKCXDMFYTFD-UHFFFAOYSA-N
storage temp.
Store at 0-8C
Purity
97%
Shipping Temperature
RT
Physical Form
White powder
Product Link
https://www.chemimpex.com/product/productinfo/5-2-4-dichlorophenyl-isoxazole-3-carboxylic-acid/16034?cid=0
Melting Point
178-186C
MSDS
https://www.chemimpex.com/product/productinfo/5-2-4-dichlorophenyl-isoxazole-3-carboxylic-acid/16034?cid=0
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.