ATEH0DDC430D
AstaTech, Inc.
(4-(PYRIDIN-3-YL)PHENYL)METHANAMINE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
[4-(3-pyridinyl)phenyl]methanamine
Inchi Code
1S/C12H12N2/c13-8-10-3-5-11(6-4-10)12-2-1-7-14-9-12/h1-7,9H,8,13H2
InChI key
FUHDBJVMRRGFTR-UHFFFAOYSA-N
Purity
95%
Country of Origin
CN
Physical Form
Pale-yellow to Yellow-brown Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=122736
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=F20167
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.