Skip to Content
Merck
All Photos(1)

Key Documents

ATE245844565

AstaTech, Inc.

2-(PYRIMIDIN-2-YLOXY)BENZOIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
216.2
MDL number:
Pricing and availability is not currently available.

Iupac Name

2-(2-pyrimidinyloxy)benzoic acid

Inchi Code

1S/C11H8N2O3/c14-10(15)8-4-1-2-5-9(8)16-11-12-6-3-7-13-11/h1-7H,(H,14,15)

InChI key

BCIHMWNOJJYBSJ-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

97%

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=1056

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=30058


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library