Skip to Content
Merck
All Photos(1)

Key Documents

ATE002883459

AstaTech, Inc.

1-N-FMOC-3-PYRROLIDINONE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
307.35
MDL number:

Iupac Name

9H-fluoren-9-ylmethyl 3-oxo-1-pyrrolidinecarboxylate

Inchi Code

1S/C19H17NO3/c21-13-9-10-20(11-13)19(22)23-12-18-16-7-3-1-5-14(16)15-6-2-4-8-17(15)18/h1-8,18H,9-12H2

InChI key

YCJGQBRHXIBEAY-UHFFFAOYSA-N

storage temp.

2-8C

Purity

97%

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=5766

Pictograms

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=58580


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library