ATE002883412
AstaTech, Inc.
2,4-DICHLORO-5-[(ETHYLTHIO)METHYL]PYRIMIDINE
About This Item
Iupac Name
2,4-dichloro-5-[(ethylsulfanyl)methyl]pyrimidine
Inchi Code
1S/C7H8Cl2N2S/c1-2-12-4-5-3-10-7(9)11-6(5)8/h3H,2,4H2,1H3
InChI key
LVMRWJLPIBPOMZ-UHFFFAOYSA-N
storage temp.
Purity
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=68391
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.