Skip to Content
Merck
All Photos(1)

Key Documents

APO465887039

Apollo Scientific Ltd

2-Bromo-3,5-difluorobenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
218
MDL number:
Pricing and availability is not currently available.

Iupac Name

2-bromo-3,5-difluorobenzonitrile

Inchi Code

1S/C7H2BrF2N/c8-7-4(3-11)1-5(9)2-6(7)10/h1-2H

InChI key

KDDKQAPQOJGHIK-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Crystals

Product Link

https://store.apolloscientific.co.uk/product/2-bromo-35-difluorobenzonitrile

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-bromo-35-difluorobenzonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library