Skip to Content
Merck
All Photos(1)

Key Documents

APO455838536

Apollo Scientific Ltd

2-Fluoro-2-methylpropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
106.1
MDL number:

Iupac Name

2-fluoro-2-methylpropanoic acid

Inchi Code

1S/C4H7FO2/c1-4(2,5)3(6)7/h1-2H3,(H,6,7)

InChI key

NZDOWZQRNZLBOY-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Low Melt

Product Link

https://store.apolloscientific.co.uk/product/2-fluoro-2-methylpropanoic-acid

Synonym

2-Fluoroisobutanoic acid, 2-Fluoro-2-methylpropionic acid, 2-Fluoroisobutyric acid

Flash Point

57-58/10mm

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-fluoro-2-methylpropanoic-acid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library