Documents
Z692891
Rota-Rack® Duo tube rack
pkg of 1 ea
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
material
polypropylene
feature
Holds 48 x 0.5 mL tubes
Holds 48 x 1.5 mL tubes
Holds 96 x 0.2 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific HS29041A
L × W × H
296 mm × 118 mm × 34 mm
General description
Work with PCR tubes right next to centrifuge tubes. The 0.2 mL tube wells are oriented in standard 96-well format to be used with PCR transfer trays.
Rotate the rack to customize for your application. Work with 96 x 0.2 mL tubes and 48 x 0.5 mL tubes or turn a module and work with 96 x 0.2 mL tubes and 48 x 1.5 mL tubes. Flip the rack and work with 48 x 0.5 mL tubes and 48 x 1.5 mL tubes. Or turn a module and work with a total of 96 x 1.5 mL tubes.
  • Tapered wells securely suspend tubes upright and makes tube removal easy
  • Imprinted alpha-numeric system makes it easy to quickly identify tubes
  • Autoclavable
Legal Information
Rota-Rack is a registered trademark of Heathrow Scientific