Z569011
Aldrich® Liebig condenser with cup
Overall H 300 mm, joint: ST/NS 24/40
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS24/40
Overall H
300 mm
Related Categories
General description
Condensate cup catches water that forms on the outer surface of the condenser during use and eliminates puddling on reactor and equipment. Tubing connects to cup′s hose barb to drain away water automatically.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC