Skip to Content
Merck
All Photos(1)

Key Documents

W246603

Sigma-Aldrich

Eucalyptus oil

natural (US), 70-75%, FCC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2466
EC Number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Eucalyptus globulus Labillardiere

Quality Level

grade

Kosher
natural (US)

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 172.510

Assay

70-75%

origin

China origin

refractive index

n20/D 1.46

bp

175 °C

density

0.909 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

herbaceous

InChI

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3

InChI key

WEEGYLXZBRQIMU-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.8 °F - closed cup

Flash Point(C)

57.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service