Skip to Content
Merck
All Photos(1)

Documents

G2001

Sigma-Aldrich

D-(+)-Gluconic acid δ-lactone

99%

Synonym(s):

δ-Gluconolactone, 1,2,3,4,5-Pentahydroxycaproic acid δ-lactone, D-(+)-Dextronic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
Beilstein:
83286
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

optical activity

[α]20/D +65°, c = 1 in H2O

mp

160 °C (dec.) (lit.)

SMILES string

OC[C@H]1OC(=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H10O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-5,7-10H,1H2/t2-,3-,4+,5-/m1/s1

InChI key

PHOQVHQSTUBQQK-SQOUGZDYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Glucono-d-lactone increased the doubling time and activated enzymes involved in the oxidative pentose phosphate pathway of Saccharomyces cerevisiae.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service