Skip to Content
Merck
All Photos(2)

Key Documents

730440

Sigma-Aldrich

Tetraacetoxy diboroxane

97%

Synonym(s):

Pyroboron tetraacetate, Tetraacetyl diborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12B2O9
CAS Number:
Molecular Weight:
273.80
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

core: boron
reagent type: catalyst

mp

147-150 °C

SMILES string

CC(=O)OB(OB(OC(C)=O)OC(C)=O)OC(C)=O

InChI

1S/C8H12B2O9/c1-5(11)15-9(16-6(2)12)19-10(17-7(3)13)18-8(4)14/h1-4H3

InChI key

ZMJHYNIVOZEJTJ-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service