Skip to Content
Merck
All Photos(1)

Key Documents

561509

Sigma-Aldrich

Benzene-D6

"100%", 99.96 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Hexadeuterobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15
Beilstein:
1905426
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.96 atom % D

Assay

≥99.0%

form

liquid

contains

0.03 % (v/v) TMS

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.497 (lit.)

bp

79.1 °C/760 mmHg (lit.)

mp

6.8 °C (lit.)

density

0.950 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

UHOVQNZJYSORNB-MZWXYZOWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzene-d6 (C6D6) is deuterated benzene that contains 0.03% (v/v) TMS (tetramethylsilane). Its Soret coefficient S(T) has been measured by transient holographic grating technique.[1] Its synthesis,13C NMR, IR and MS spectra have been reported.[2][3]

Application

Benzene-d6 (C6D6) may be used in the following:
  • Synthesis of isotopically labeled samples.[4]
  • As a 1H and 13C-NMR solvent.[5][6]
  • In the internal calibration of benzene stock solution.[7]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Spectral Analysis of Benzene-d6.
Guo YZ, et al.
Advanced Materials Research, 1061-1062, 301-306 (2014)
Biomonitoring of benzene and toluene in human blood by headspace-solid-phase microextraction.
Schimming E, et al.
Fresenius Journal of Analytical Chemistry, 363(1), 88-91 (1999)
Surprising Titanium complexes bearing η2-pyrazolato ligands: synthesis, structure, and molecular orbital studies.
Guzei IA, et al.
Journal of the American Chemical Society, 119(14), 3387-3388 (1997)
Structure-property relationships in nematic gold nanoparticles.
Cseh L and Mehl GH.
Journal of Materials Chemistry, 17(4), 311-315 (2007)
Infrared intensities of liquids XXII: Optical and dielectric constants, molar polarizabilities, and integrated intensities of liquid benzene-d6 at 25?C between 5000 and 450 cm-1.
Bertie JE and Keefe CD.
Fresenius Journal of Analytical Chemistry, 362(1), 91-108 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service