Skip to Content
Merck
All Photos(1)

Key Documents

516511

Sigma-Aldrich

Di-tert-butyl polysulfide

Synonym(s):

TPS® 44

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:

Legal Information

Product of Arkema
TPS is a registered trademark of Arkema France

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service