Skip to Content
Merck
All Photos(1)

Key Documents

462535

Sigma-Aldrich

Polybutadiene-graft-poly(methyl acrylate-co-acrylonitrile)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

density

1.15 g/mL at 25 °C (lit.)

SMILES string

C=CC=C.C=CC#N.COC(=O)C=C

Application

Packaging containers.

Features and Benefits

Good gas barrier properties, excellent chemical resistance, high film clarity and scratch resistance.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service