Skip to Content
Merck
All Photos(1)

Documents

458260

Sigma-Aldrich

2,4,6-Triphenylborazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18B3N3
CAS Number:
Molecular Weight:
308.79
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

reagent type: catalyst
core: boron

mp

180 °C (dec.) (lit.)

SMILES string

c1ccc(cc1)-b2[nH]b([nH]b([nH]2)-c3ccccc3)-c4ccccc4

InChI

1S/C18H18B3N3/c1-4-10-16(11-5-1)19-22-20(17-12-6-2-7-13-17)24-21(23-19)18-14-8-3-9-15-18/h1-15,22-24H

InChI key

QYWXVASQMOFUGX-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service