Skip to Content
Merck
All Photos(1)

Key Documents

39785

Sigma-Aldrich

6-Chloro-1-hydroxybenzotriazole dihydrate

≥98.0% (HPLC)

Synonym(s):

Cl-HOBt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClN3O · 2H2O
CAS Number:
Molecular Weight:
205.60
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98.0% (HPLC)

impurities

~17.5% water

mp

190-195 °C (dec.)

SMILES string

O.O.On1nnc2ccc(Cl)cc12

Caution

For safe storage and transport the material was desensitized with two equivalents of water

Other Notes

Reagent to suppress racemization in peptide coupling, which is not toxic.[1]

Pictograms

Exploding BombExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Expl. 1.3

Storage Class Code

1 - Explosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spec. Chem., July/August, 2-2 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service