Skip to Content
Merck
All Photos(1)

Documents

377643

Sigma-Aldrich

3-Cyanopropyltrichlorosilane

97%

Synonym(s):

4-(Trichlorosilyl)butyronitrile, Trichloro(3-cyanopropyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3Si(CH2)3CN
CAS Number:
Molecular Weight:
202.54
Beilstein:
906924
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.463 (lit.)

bp

237-238 °C (lit.)

density

1.3 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)CCCC#N

InChI

1S/C4H6Cl3NSi/c5-9(6,7)4-2-1-3-8/h1-2,4H2

InChI key

HMFFOEBLYHLRQN-UHFFFAOYSA-N

Application

3-Cyanopropyltrichlorosilane may be used to functionalize multiwalled carbon nanotubes to prevent aggregation in solutions.{37}

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of an adherent polyacrylonitrile/carbon nanotubes composite film onto a polyacrylonitrile brush electrografted on copper
Vast L, et al.
Journal of Nanoscience and Nanotechnology, 7(10), 3404-3410 (2007)
Behaviour of synthetic corticoids in ointment on 3-cyanopropyltrichlorosilane in high-performance thin-layer chromatography.
M Okamoto et al.
Journal of chromatography, 369(2), 403-407 (1986-11-21)
[Evaluation of 3-cyanopropyltrichlorosilane treated thin layer chromatographic plates. Application to analysis of paeoniflorin in paeony roots].
M Okamoto et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 106(9), 837-839 (1986-09-01)
Supports for reverse-phase high-performance liquid chromatography of large proteins.
R V Lewis et al.
Analytical biochemistry, 104(1), 153-159 (1980-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service