Skip to Content
Merck
All Photos(1)

Documents

289450

Sigma-Aldrich

4-Fluoro-1-naphthoic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC10H6CO2H
CAS Number:
Molecular Weight:
190.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

224-227 °C (lit.)

SMILES string

OC(=O)c1ccc(F)c2ccccc12

InChI

1S/C11H7FO2/c12-10-6-5-9(11(13)14)7-3-1-2-4-8(7)10/h1-6H,(H,13,14)

InChI key

DEWIOKQDRWFLFW-UHFFFAOYSA-N

Application

4-Fluoro-1-naphthoic acid is an inhibitor of Yersinia PTP YopH. It contains an extended aromatic system and its IC50 value was measured.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fubo Liang et al.
The Journal of biological chemistry, 278(43), 41734-41741 (2003-07-31)
Yersinia are causative agents in human diseases ranging from gastrointestinal syndromes to Bubonic Plague. There is increasing risk of misuse of infectious agents, such as Yersinia pestis, as weapons of terror as well as instruments of warfare for mass destruction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service