Skip to Content
Merck
All Photos(1)

Key Documents

101443

Sigma-Aldrich

Agmatine sulfate salt

Enzymatic decarboxylation product of arginine., 97%

Synonym(s):

1-Amino-4-guanidinobutane sulfate salt, 4-Guanidinobutylamine sulfate salt, N-(4-Aminobutyl)guanidine sulfate salt

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
H2N(CH2)4NHC(=NH)NH2·H2SO4
CAS Number:
Molecular Weight:
228.27
Beilstein:
3918807
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

mp

234-238 °C (lit.)

SMILES string

OS(O)(=O)=O.NCCCCNC(N)=N

InChI

1S/C5H14N4.H2O4S/c6-3-1-2-4-9-5(7)8;1-5(2,3)4/h1-4,6H2,(H4,7,8,9);(H2,1,2,3,4)

InChI key

PTAYFGHRDOMJGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Putative endogenous neurotransmitter at imidazoline receptors; displaces clonidine at α2-adrenergic and at imidazoline receptors; blocks NMDA-activated ion channels in hippocampal neurons.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D S El-Agamy et al.
Human & experimental toxicology, 33(6), 650-660 (2014-02-08)
There is a large body of evidence that nitric oxide (NO) formation is implicated in mediating silica-induced pulmonary fibrosis. As a reactive free radical, NO may not only contribute to lung parenchymal tissue injury but also has the ability to
Julia Torres et al.
Organic & biomolecular chemistry, 13(27), 7500-7512 (2015-06-13)
Within all the eukaryotic cells myo-inositol phosphates (InsPs) are an important group of biomolecules that are potentially related to signaling functions. The most abundant member of this family in nature is InsP6 (phytate, L(12-) in its fully deprotonated form). The
Patrícia Pereira et al.
Journal of chromatography. A, 1368, 173-182 (2014-12-03)
MicroRNA-based therapeutic applications have fostered a growing interest in the development of microRNAs purification processes in order to obtain the final product with high purity degree, good quality and biologically active. The pre-miR-29 deficiency or overexpression has been associated to
Ling Li et al.
Molecular cell, 80(3), 525-540 (2020-10-18)
Well-balanced and timed metabolism is essential for making a high-quality egg. However, the metabolic framework that supports oocyte development remains poorly understood. Here, we obtained the temporal metabolome profiles of mouse oocytes during in vivo maturation by isolating large number of
Ângela Sousa et al.
Journal of chromatography. A, 1366, 110-119 (2014-09-30)
Histamine and agmatine amino acid derivatives were immobilized into monolithic disks, in order to combine the specificity and selectivity of the ligand with the high mass transfer and binding capacity offered by monolithic supports, to purify potential plasmid DNA biopharmaceuticals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service