AABH97CE8908
AA BLOCKS, INC.
2-[(3,4-dimethylphenoxy)methyl]benzoic acid
About This Item
Iupac Name
2-[(3,4-dimethylphenoxy)methyl]benzoic acid
Inchi Code
1S/C16H16O3/c1-11-7-8-14(9-12(11)2)19-10-13-5-3-4-6-15(13)16(17)18/h3-9H,10H2,1-2H3,(H,17,18)
InChI key
RSUJHHWCAZHBFN-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Product Link
Synonym
MSDS
https://www.aablocks.com/downloadSds?cas=77389-77-6
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.