Merck
  • Isoguanosine substitution of conserved adenosines in the hammerhead ribozyme.
Isoguanosine substitution of conserved adenosines in the hammerhead ribozyme.
Biochemistry (1994-10-11)
M M Ng, F Benseler, T Tuschl, F Eckstein
ABSTRACT
Isoguanosine has been incorporated into a 34-mer hammerhead ribozyme by the solid-phase phosphoramidite method, using an acetamidine base protecting group. The activity of the hammerhead ribozyme when singly mutated to isoguanosine at the adenosine positions 6, 9, and 13 was 1-2-fold less than the wild-type activity. Mutations to 2-aminopurine ribonucleoside at positions 9 and 13 were 5-fold reduced in activity, but that at position 6 was approximately 30-fold reduced. These results support the view that the 6-amino functions of A6, A9, and A13 are not very important for catalysis. The 2-position of A6 tolerates a carbonyl function but not an amino group, whereas A9 and A13 tolerate both functional groups. The tolerance of a 2-amino group at A9 and A13 makes G(anti)/A(anti) Watson-Crick type base mispairing for G12/A9 and A13/G8 unlikely.
Sigma-Aldrich
Acetamidine hydrochloride, 95%