Skip to Content
Merck
All Photos(1)

Key Documents

218146

Sigma-Aldrich

2-Chloro-6-fluorophenylacetonitrile

98%

Synonym(s):

2-Chloro-6-fluorobenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)CH2CN
CAS Number:
Molecular Weight:
169.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

41-42 °C (lit.)

functional group

chloro
fluoro
nitrile

SMILES string

Fc1cccc(Cl)c1CC#N

InChI

1S/C8H5ClFN/c9-7-2-1-3-8(10)6(7)4-5-11/h1-3H,4H2

InChI key

ZGSAFMIRVLOISC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-6-fluorophenylacetonitrile may be used in synthesis of aryl fluorinated building blocks.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service