Skip to Content
Merck

SML3105

K-7174 dihydrochloride

≥98% (HPLC)

Synonym(s):

1,4-bis((E)-5-(3,4,5-Trimethoxyphenyl)pent-4-enyl)-1,4-diazepane dihydrochloride, K 7174 dihydrochloride, K7174 dihydrochloride, N,N?-bis-(E)-[5-(3,4,5-Trimethoxyphenyl)-4-pentenyl]homopiperazine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C33H48N2O6 · 2HCl
CAS Number:
Molecular Weight:
641.67
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

COC(C(OC)=C1)=C(C=C1/C=C/CCCN(CCC2)CCN2CCC/C=C/C3=CC(OC)=C(C(OC)=C3)OC)OC.[H]Cl.[H]Cl

InChI key

JKAQFPBRMVEHBD-CHBZAFCASA-N

Biochem/physiol Actions

K-7174 is a homopiperazine with dual GATA and proteasome inhibitory activtiy. K-7174 selective inhibts GATA-mediated gene regulations in cultures (10-30 μM; VCAM-1 expression in HUVECs or Epo suppression in Hep3B cells) and in vivo (30 mg/kg in mice via i.p. against IL-1beta- or TNF-alpha-induced anemia). K-7174 exhibits anti-myeloma activity in vitro (10-25 μM) and in vivo (50 mg/kg/day p.o. in mice) by targeting the active pockets of β1, β2 and β5 subunits of proteasome along hydrophobic grooves in the direction of the β7, β1 and β4 subunits in a manner distinct from that of bortezomib.
Orally active, dual GATA and proteasome inhibitor in vitro and in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library