Skip to Content
Merck
All Photos(1)

Documents

S1814.PB5

85011402, human embryo, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

S1814.PB5, 85011402

biological source

human embryo

growth mode

Adherent

karyotype

Diploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human embryo fibroblast

Cell Line Description

Human embryo fibroblast cell line.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 10,12
D16S539: 11
D5S818: 13
D7S820: 10,11
THO1: 7,9
TPOX: 9,11
vWA: 15

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS). Cells should also grow in DMEM or RPMI 1640. This cell line also grow well in HAT.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 i.e. seeding 3-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service