F0120010
Flecainide impurity B
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-(Aminomethyl)piperidine, (RS)-1-(Piperidin-2-yl)methanamine, 2-Piperidinemethanamine
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
flecainide
manufacturer/tradename
EDQM
refractive index
n20/D 1.4854
density
0.9406 g/mL at 25 °C
application(s)
pharmaceutical (small molecule)
format
neat
shipped in
dry ice
storage temp.
−20°C
SMILES string
NCC1CCCCN1
InChI
1S/C6H14N2/c7-5-6-3-1-2-4-8-6/h6,8H,1-5,7H2
InChI key
RHPBLLCTOLJFPH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Reactant for synthesis of:
- Annulated N-heterocyclic carbene ligands
- M3 Muscarinic receptor antagonists
- Selective 5-ht5A receptor antagonists
- Ternary platinum(II) complexes
Packaging
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
154.9 °F
Flash Point(C)
68.3 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service