Skip to Content
Merck
All Photos(2)

Key Documents

MABF1975

Sigma-Aldrich

Anti-Herpes Simplex Virus Glycoprotein D Antibody, clone LP14

clone LP14, from mouse

Synonym(s):

HSV glycoprotein D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
LP14, monoclonal
application:
ICC
WB
inhibition assay
species reactivity:
virus
technique(s):
immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LP14, monoclonal

species reactivity

virus

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

Herpes Simplex Virus envelope Glycoprotein D (UniProt: Q69091) is also known as gD. The envelope of herpes simplex virus (HSV) is complex and contains at least 10 virus-encoded glycoproteins, some of which mediate virus entry into the host cell. HSV virus enters the cell by first fusing its envelope with a membrane of the host cell. Five viral envelope glycoproteins are reported to be involved in this entry process. Binding of glycoprotein D of HSV to a cell surface receptor is required to trigger membrane fusion during entry into host cells. Glycoproteins C and B promote attachment by interacting with cell surface proteoglycans and then glycoprotein D binds to the herpes virus entry mediator (HVEM) receptor that initiates a process that ultimately leads to glycoprotein B-mediated membrane fusion. Glycoprotein D is synthesized with a signal peptide (aa 1-25) that is subsequently cleaved off. It is a homodimeric protein that contains two zinc-binding sites (aa 64 and 264). The ectodomain region of glycoprotein D contains three sites for the addition of N-linked oligosaccharides (N-CHO) and six cysteine residues arranged into three disulfide bonds. During virion morphogenesis, glycoprotein D probably accumulates in the endosomes and trans-Golgi where secondary envelopment occurs. (Ref.: Whitbeck JC et al. (1997). J. Virol. 71(8); p. 6083 6093; Nicola, AV et al. (1998). J Virol. 72(5): 3595-3601; Giovine, PD et al. (2011). PLos Pathog. 7(9); e1002277).

Specificity

Clone LP14 detects glycoprotein D of Herpes Simplex virus.

Immunogen

Whole Herpex Simplex Virus 2 (HSV-2).

Application

Anti-Herpes Simplex Virus Glycoprotein D, clone LP14, Cat. No. MABF1975, is a mouse monoclonal antibody that detects Glycoprotein D of Herpes Simplex Virus and has been tested for use in Inhibition, Immunocytochemistry, and Western Blotting.
Research Category
Inflammation & Immunology
Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected Herpes Simplex Virus Glycoprotein D in lysates from VERO cells transfected with HSV-1.

Western Blotting Analysis: A representative lot detected Herpes Simplex Virus Glycoprotein D in Western Blotting applications (Lau, S.Y., et. al. (2015). Viruses. 7(3):915-38; Zenner, H.L., et. al. (2011). J Virol. 85(16):8012-21; Pawliczek, T., et. al. (2009). J Virol. 83(21):11254-64).

Western Blotting Analysis: A representative lot detected Herpes Simplex Virus Glycoprotein D in Western Blotting applications (Ren, Y., et. al. (2012). J Gen Virol. 93(Pt 2):319-29).

Immunocytochemistry Analysis: A representative lot detected Herpes Simplex Virus Glycoprotein D in Immunocytochemistry applications (Lau, S.Y., et. al. (2015). Viruses. 7(3):915-38).

Inhibition Analysis: A representative lot inhibited the fusion of cells infected with HSV-1. (Minson, A.C., et. al. (1986). J Gen Virol. 67(6); 1001-1013.

Quality

Evaluated by Western Blotting in HEK293T cell lysate transfected with HSV gD.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected Herpes Simplex Virus Glycoprotein D in HEK293T cell lysate transfected with HSV gD.

Target description

~50 and 55 kDa observed; 43.35 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harrison M McCann et al.
ACS chemical biology, 17(5), 1269-1281 (2022-05-07)
Covalent antibody recruiting molecules (cARMs) constitute a proximity-inducing chemical strategy to modulate the recognition and elimination of cancer cells by the immune system. Recognition is achieved through synthetic bifunctional molecules that use covalency to stably bridge endogenous hapten-specific antibodies like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service