682160-M
Xestospongin C, Xestospongia sp.
An oxaquinolizidine alkaloid isolated from a marine sponge.
Synonym(s):
Xestospongin C, Xestospongia sp., XeC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
film
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white to off-white
solubility
DMSO: 1 mg/mL
ethanol: 1 mg/mL
methanol: 1 mg/mL
storage temp.
−20°C
General description
A macrocyclic bis-1-oxaquinolizidine natural product isolated from a marine sponge. A very potent, reversible and membrane-permeable blocker of IP3-mediated Ca2+ release (IC50 = 358 nM) that does not interact with the IP3-binding site. Displays high selectivity over the skeletal isoform of the ryanodine receptor type 1 (Ry1R). Also blocks bradykinin- and carbamylcholine-induced Ca2+ efflux from the ER stores in a reversible manner.
An oxaquinolizidine alkaloid isolated from a marine sponge. A very potent, reversible, and membrane-permeable blocker of IP3-mediated Ca2+ release (IC50 = 358 nM) that does not interact with the IP3-binding site. Displays high selectivity over the skeletal isoform of the ryanodine receptor type 1 (RyR-1). Also blocks bradykinin- and carbamylcholine-induced Ca2+ efflux from the endoplasmic reticulum stores in a reversible manner.
Biochem/physiol Actions
Primary Target
IP3-mediated Ca2+ release
IP3-mediated Ca2+ release
Target IC50: 358 nM in blocking IP3-mediated Ca2+ release
Warning
Toxicity: Harmful (C)
Preparation Note
Sonication may be required to achieve complete solubilization.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Oka, T., et al. 2002. Br. J. Pharmacol.135, 1959.
Gafni, J., et al. 1997. Neuron 19, 723.
Tinker, A., and Williams, A.J. 1995. Biophys. J. 68, 111.
Callaway, C., et al. 1994. J. Biol. Chem. 269, 15876.
Quirion, J.C., et al. 1992. J. Nat. Prod. 55, 1505.
Nakagawa, M., et al. 1984. Tetrahedron Lett.25, 3227.
Gafni, J., et al. 1997. Neuron 19, 723.
Tinker, A., and Williams, A.J. 1995. Biophys. J. 68, 111.
Callaway, C., et al. 1994. J. Biol. Chem. 269, 15876.
Quirion, J.C., et al. 1992. J. Nat. Prod. 55, 1505.
Nakagawa, M., et al. 1984. Tetrahedron Lett.25, 3227.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service