MilliporeSigma
  • Whole blood assay for elastase, chymotrypsin, matrix metalloproteinase-2, and matrix metalloproteinase-9 activity.
Whole blood assay for elastase, chymotrypsin, matrix metalloproteinase-2, and matrix metalloproteinase-9 activity.
Analytical chemistry (2010-09-11)
Roy B Lefkowitz, Geert W Schmid-Schönbein, Michael J Heller
ABSTRACT
The ability to measure protease activity in the blood is important for the development of future diagnostics and for biomedical research. Presently, protease assays require sample preparation, making them time-consuming, costly, less accurate, and unsuitable for point-of-care (POC) diagnostics. Recently, we demonstrated a unique method for measuring clinically relevant levels of trypsin activity in only a few microliters of whole blood. This assay utilizes a charge-changing fluorescent peptide substrate that produces a positively charged fluorescent product fragment upon cleavage by the target protease. Using a simple electrophoretic format, the fragments could be rapidly separated, concentrated, and detected directly from a whole blood sample. We now report on the development of new protease substrates for the measurement of elastase, chymotrypsin, matrix metalloproteinase (MMP)-2, and MMP-9 activity in whole blood. In these studies, detection limits ranging from 1 to 40 pg in 6 μL of 1× phosphate-buffered saline (PBS) (0.2-6 ng/mL) were achieved after a only 1 h reaction of enzyme and substrate. In subsequent experiments measuring spiked protease in whole blood (with endogenous protease present), detection limits ranging from 100 to 200 ng/mL were achieved after a 1 h reaction. Thus, these new substrates demonstrate broad applicability toward clinically relevant detection of important disease-relevant proteases.
Sigma-Aldrich
α1-Antitrypsin from human plasma, salt-free, lyophilized powder