Merck
Jellyfish Stings: A Practical Approach.
Wilderness & environmental medicine (2015-05-04)
Najla A Lakkis, Grace J Maalouf, Dina M Mahmassani
ABSTRACT
Jellyfish have a worldwide distribution. Their stings can cause different reactions, ranging from cutaneous, localized, and self-limited to serious systemic or fatal ones, depending on the envenoming species. Several first aid treatments are used to manage such stings but few have evidence behind their use. This review of the literature describes and discusses the different related first aid and treatment recommendations, ending with a summarized practical approach. Further randomized controlled trials in this field are needed.
Sigma-Aldrich
Aluminum sulfate, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Aluminum sulfate, meets USP testing specifications