
Glucosamine Impurity B

Synonym(s):
Fructosazine, 2,5-bis(D-Arabinotetrahydroxybutyl)pyrazine
Empirical Formula (Hill Notation):
C12H20N2O8
CAS Number:
Molecular Weight:
320.30
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing