Skip to Content
Merck
All Photos(1)

Key Documents

27016

Supelco

Aluminum crimp seal (without septum), silver aluminum

tear-away, diam. 20 mm, pkg of 100 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
Pricing and availability is not currently available.

material

silver aluminum

feature

tear-away

packaging

pkg of 100 ea

O.D.

20 mm

diam.

20 mm

compatibility

for use with 20 mm crimp seal vial

Looking for similar products? Visit Product Comparison Guide

Application

It was used for sealing 100 mL serum bottles fitted with gray stopper during analysis of biogas emission in lagoons.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lagoon-biogas emissions and carbon balance estimates of a swine production facility.
DeSutter, Tom M., and Jay M. Ham.
Journal of Environmental Quality, 34.1, 198-206 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service