Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100624

Sigma-Aldrich

Anti-DPYSL2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CRMP2, Anti-DHPRP2, Anti-DRP2, Anti-Dihydropyrimidinase-like 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

62 kDa

species reactivity

dog, human, guinea pig, bovine, horse, mouse, rat, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPYSL2(1808)

Immunogen

Synthetic peptide directed towards the middle region of human DPYSL2

Biochem/physiol Actions

DPYSL2 belongs to the DHOase family. It is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. The protein plays a role in axon guidance, neuronal growth cone collapse and cell migration.

Sequence

Synthetic peptide located within the following region: NIFEGMECRGSPLVVISQGKIVLEDGTLHVTEGSGRYIPRKPFPDFVYKR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service