94508
3-[N,N-Dimethyl(3-palmitoylaminopropyl)ammonio]-propanesulfonate
Synonym(s):
3-[N,N-Dimethyl-N-(3-palmitamidopropyl)ammonio]propane-1-sulfonate, Amidosulfobetain-16
About This Item
Recommended Products
description
zwitterionic
form
powder
mol wt
462.73 g/mol
technique(s)
electrophoresis: suitable
impurities
≤5.0% water
solubility
H2O: 50 mg/mL, faintly turbid, faintly yellow (hot)
application(s)
detection
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O
InChI
1S/C24H50N2O4S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-19-24(27)25-20-17-21-26(2,3)22-18-23-31(28,29)30/h4-23H2,1-3H3,(H-,25,27,28,29,30)
InChI key
CLCSYZQBLQDRQU-UHFFFAOYSA-N
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service