Skip to Content
Merck
All Photos(1)

Key Documents

371818

Sigma-Aldrich

G-Protein βγ Binding Peptide, mSIRK

A cell-permeable, N-myristoylated G-protein βγ selective binding peptide that disrupts the interaction between α and βγ subunits and promotes the α subunit dissociation without stimulating nucleotide exchange.

Synonym(s):

G-Protein βγ Binding Peptide, mSIRK, mSIRK, myr-SIRKALNILGYPDYD-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C93H150N20O25
Molecular Weight:
1948.31
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable, N-myristoylated G-protein βγ selective binding peptide that disrupts the interaction between α and βγ subunits and promotes the α subunit dissociation without stimulating nucleotide exchange. Shown to cause a rapid activation of ERK1/2 (EC50 ~ 2.5 - 5 µM) in rat arterial smooth muscle and Rat2 cells. Also promotes the phosphorylation of JNK and p38 MAP kinase, phospholipase C and causes Ca2+ release from internal stores. Negative control is also available (Cat. No. 371820).

Biochem/physiol Actions

Cell permeable: yes
EC50 ~ 2.5 - 5 µM in activation of ERK1/2 in rat arterial smooth muscle and Rat2 cells
Primary Target
N-myristoylated G-protein βγ selective binding
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Myr-N-Ser-Ile-Arg-Lys-Ala-Leu-Asn-Ile-Leu-Gly-Tyr-Pro-Asp-Tyr-Asp-OH

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Wei Zhang et al.
Molecular medicine reports, 21(2), 720-730 (2020-01-25)
In the present study, the mechanism by which carboxyl terminal activating region 3 (CTAR3) of latent membrane protein 1 (LMP1), encoded by the Epstein‑Barr virus, regulated cell proliferation and protein expression was investigated in the nasopharyngeal epithelial cell line NP69.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service